missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC22A6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526244
This item is not returnable.
View return policy
Description
SLC22A6 Polyclonal Antibody for Western Blot, IHC (P), Flow
The protein is involved in the sodium-dependent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and may be localized to the basolateral membrane.
Specifications
| SLC22A6 | |
| Polyclonal | |
| Unconjugated | |
| SLC22A6 | |
| hOAT1; hPAHT; hROAT1; hypothetical protein zgc:77073; kidney-specific transport protein; mNKT; mOat1; mROAT1; NKT; Novel kidney transcript; oat; OAT1; oat3; Orctl1; Organic anion transporter 1; organic cationic transporter-like 1; PAH transporter; PAHT; para-aminohippurate transporter; rbROAT1; Renal organic anion transporter 1; renal organic anion transporter 1 (rbOAT1); ROAT1; rROAT1; slc22a6; slc22a6l; solute carrier family 22 (organic anion transporter), member 6; solute carrier family 22 (organic anion transporter), member 6, like; solute carrier family 22 member 6; solute carrier family 22 member 6 isoform b; solute carrier family 22 member 6, like; solute carrier family 22, member 6; zgc:77073 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 9356 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q4U2R8 | |
| SLC22A6 | |
| KLH conjugated synthetic peptide between 513-541 amino acids from the C-terminal region of human SLC22A6. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction