missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC22A5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537389
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 63 kDa. Purity is >95% by SDS-PAGE.
Carnitine (b-hydroxy-g-trimethylaminobutyrate) is a small, highly polar compound that aids in the b-oxidation of long-chain fatty acids. Organic cation/carnitine transporters (OCTN) assist in the elimination of cationic compounds, including xenobiotics, and transport carnitine for reabsorption in the kidney. Similar to organic cation transporters (OCT), OCTN proteins localize to the plasma membrane of epithelial cells. OCTN1 is expressed in kidney, trachea, bone marrow and fetal liver. OCTN2 is abundantly expressed in kidney, skeletal muscle, placenta and heart. OCTN3 is strongly expressed in testis and weakly expressed in kidney. The gene encoding human OCTN1 maps to chromosome 5 and the gene encoding human OCTN2 maps to chromosome 5q31. Mutations in the gene encoding OCTN2 leads to systemic carnitine deficiency (SCD), an autosomal recessive disorder characterized by cardiomyopathy, skeletal myopathy, lethargy, hypoglycemia and hyperammonemia.
Specifications
| SLC22A5 | |
| Polyclonal | |
| Unconjugated | |
| SLC22A5 | |
| CDSP; CT1; high-affinity carnitine transporter; high-affinity sodium dependent carnitine cotransporter; high-affinity sodium-dependent carnitine cotransporter; integral membrane transport protein; juvenile visceral steatosis; jvs; Lstpl; Octn2; OCTN2VT; Organic cation/carnitine transporter 2; S22A5; SLC22A5; solute carrier family 22 (organic cation transporter), member 5; solute carrier family 22 (organic cation/carnitine transporter), member 5; solute carrier family 22 member 5; solute carrier family 22, member 5; UST2r | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20520, 29726, 6584 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O70594, O76082, Q9Z0E8 | |
| SLC22A5 | |
| Recombinant full length human SLC22A5. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction