missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC22A4 Monoclonal Antibody (C1), Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

£177.00 - £385.00

Specifications

Antigen SLC22A4
Clone C1
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17684026
View Documents
Invitrogen™
MA543572
20 μL
£177.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17674026
GREENER_CHOICE
View Documents
Invitrogen™
MA543571
100 μL
£385.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Carnitine (b-hydroxy-g-trimethylaminobutyrate) is a small, highly polar compound that aids in the b-oxidation of long-chain fatty acids. Organic cation/carnitine transporters (OCTN) assist in the elimination of cationic compounds, including xenobiotics, and transport carnitine for reabsorption in the kidney. Similar to organic cation transporters (OCT), OCTN proteins localize to the plasma membrane of epithelial cells. OCTN1 is expressed in kidney, trachea, bone marrow and fetal liver. OCTN2 is abundantly expressed in kidney, skeletal muscle, placenta and heart. OCTN3 is strongly expressed in testis and weakly expressed in kidney. The gene encoding human OCTN1 maps to chromosome 5 and the gene encoding human OCTN2 maps to chromosome 5q31. Mutations in the gene encoding OCTN2 leads to systemic carnitine deficiency (SCD), an autosomal recessive disorder characterized by cardiomyopathy, skeletal myopathy, lethargy, hypoglycemia and hyperammonemia.
Specifications

Specifications

SLC22A4
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
ergothioneine transporter; ET transporter; ETT; integral membrane transport protein; LSTP-Like 2; Octn1; organic cation transporter 1; organic cation transporter OCTN1; organic cation/carnitine transporter 1; SLC22A4; solute carrier family (organic cation transporter), member 4; solute carrier family 22 (organic cation transporter), member 4; solute carrier family 22 (organic cation/ergothioneine transporter), member 4; solute carrier family 22 (organic cation/zwitterion transporter), member 4; solute carrier family 22 member 4; UT2H
SLC22A4
IgG1 κ
Protein A/G
Antibody
C1
Western Blot
Unconjugated
Mouse
Human
Q9H015
6583
Leu42-Lys141 (Accession Q9H015), with N-terminal His Tag.
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
SLC22A4
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.