missing translation for 'onlineSavingsMsg'
Learn More

SLC22A3 Rabbit anti-Human, Mouse, Clone: 1K0A4, Novus Biologicals™

Product Code. 18350413 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350413 100 μg 100µL
18331356 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18350413 Supplier Novus Biologicals Supplier No. NBP315897100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SLC22A3 Monoclonal antibody specifically detects SLC22A3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1K0A4
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EMTEMTH, Extraneuronal monoamine transporter, OCT3EMT organic cation transporter 3, Organic cation transporter 3, solute carrier family 22 (extraneuronal monoamine transporter), member 3, solute carrier family 22 member 3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 301-450 of human SLC22A3 (O75751). GDKALQILRRIAKCNGKYLSSNYSEITVTDEEVSNPSFLDLVRTPQMRKCTLILMFAWFTSAVVYQGLVMRLGIIgGNLYIDFFISGVVELPGALLILLTIERLGRRLPFAASNIVAGVACLVTAFLPEGIAWLRTTVATLGRLGITMAF
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6581
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.