missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC22A25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113010
This item is not returnable.
View return policy
Description
SLC22A25 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
SLC22A25 gene ontology annotations related to this gene include transmembrane transport.
Specifications
| SLC22A25 | |
| Polyclonal | |
| Unconjugated | |
| SLC22A25 | |
| HIMTP; integral membrane transport UST1r; MGI:2442751, MGI:2385316, MGI:3042283, MGI:3645714, MGI:3605624, MGI:2442750; organic anion transporter UST6; putative integral membrane transport UST1r; putative UST1-like organic anion transporter; SLC22A25; Slc22a9; solute carrier family 22 (organic anion/cation transporter), member 9; solute carrier family 22 member 24; solute carrier family 22 member 25; solute carrier family 22, member 25; Ust1; Ust1r; UST6 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 387601 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3.44 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q6T423 | |
| SLC22A25 | |
| Recombinant Human Solute carrier family 22 member 25 protein (31-145AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction