missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC22A25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571031
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 85%; Dog: 92%; Horse: 92%; Human: 100%; Mouse: 83%; Pig: 100%; Rabbit: 93%; Rat: 83%.
SLC22A25 gene ontology annotations related to this gene include transmembrane transport.
Specifications
| SLC22A25 | |
| Polyclonal | |
| Unconjugated | |
| SLC22A25 | |
| HIMTP; integral membrane transport UST1r; MGI:2442751, MGI:2385316, MGI:3042283, MGI:3645714, MGI:3605624, MGI:2442750; organic anion transporter UST6; putative integral membrane transport UST1r; putative UST1-like organic anion transporter; SLC22A25; Slc22a9; solute carrier family 22 (organic anion/cation transporter), member 9; solute carrier family 22 member 24; solute carrier family 22 member 25; solute carrier family 22, member 25; Ust1; Ust1r; UST6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 387601 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6T423 | |
| SLC22A25 | |
| synthetic peptide directed towards the middle region of human SLC22A25. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction