missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLC22A2/OCT2 Polyclonal specifically detects SLC22A2/OCT2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Single Cell Western.
Specifications
Specifications
| Antigen | SLC22A2/OCT2 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot Reactivity reported in scientific literature (PMID: 26045896)., Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000, Single Cell Western Single Cell Western reported by an internal validation at a 1:20 dilution |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | O15244 |
| Gene Alias | hOCT2, MGC32628, OCT2Organic cation transporter 2, solute carrier family 22 (organic cation transporter), member 2, solute carrier family 22 member 2 |
| Gene Symbols | SLC22A2 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?