missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC22A18AS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564365
This item is not returnable.
View return policy
Description
Immunogen sequence: ELPGSEGMWE NCPLGWVKKK ASGTLAPLDF LLQRKRLWLW ASEPVRPQPQ GIHRFREARR QFCRMRGSRL TGGRKG Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 28%.
The SLC22A18/SLC22A18AS genes are a sense-antisense pair located at human chromosome segment 11p15.5. These genes are paternally imprinted: paternal alleles are silenced and maternal alleles are expressed. SLC22A18AS (antisense) is associated to SLC22A18 (sense). These two genes share, in divergent orientations, 31 bp in their 5' regions (between the first exon of SLC22A18AS and the second exon of SLC22A18).
Specifications
| SLC22A18AS | |
| Polyclonal | |
| Unconjugated | |
| SLC22A18AS | |
| Beckwith-Wiedemann region 1B; beckwith-Wiedemann syndrome chromosomal region 1 candidate gene B protein; Beckwith-Wiedemann syndrome chromosome region 1, candidate b; BWR1B; BWSCR1B; ORCTL2S; organic cation transporter-like 2 antisense; organic cation transporter-like protein 2 antisense protein; p27-Beckwith-Wiedemann region 1 B; p27-BWR1B; SLC22A18AS; SLC22A1LS; solute carrier family 22 (organic cation transporter), member 18 antisense; solute carrier family 22 (organic cation transporter), member 1-like antisense; solute carrier family 22 member 18 antisense; solute carrier family 22 member 18 antisense protein; solute carrier family 22 member 1-like antisense protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5003 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N1D0 | |
| SLC22A18AS | |
| Recombinant protein corresponding to Human SLC22A18AS. Recombinant protein control fragment (Product #RP-103667). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction