missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC22A17 Polyclonal Antibody

Description
A suggested positive control is SK-N-SH cell lysate. PA5-20543 can be used with blocking peptide PEP-0663.
The Slc22 family of organic anion and cation transporters (OATs, OCTs, OCTNs) are transmembrane proteins expressed predominantly in kidney and liver. Each contain 12 predicted alpha-helical transmembrane domains (TMDs) and one large extracellular loop between TMDs 1 and 2. Transporters of the SLC22 family function in different ways such as uniporters that mediate facilitated diffusion in either direction (OCTs), as anion exchangers (OAT1, OAT3 and URAT1), and as Na(+)/l-carnitine cotransporter (OCTN2). Slc22 family members participate in the absorption and/or excretion of drugs, xenobiotics, and endogenous compounds in intestine, liver, and kidney, and perform homeostatic functions in brain and heart. Mutations in the Slc22 family may cause specific diseases such as primary systemic carnitine deficiency or idiopathic renal hypouricemia and may change drug absorption or excretion. Recent studies show the expression of Slc22A17 as receptor for Lipocalin 2 is relatively high in hematopoietic stem cells.
Specifications
Specifications
| Antigen | SLC22A17 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SLC22A17 |
| Gene Accession No. | Q8WUG5, Q9D9E0, Q9P290 |
| Gene Alias | 1700094C23Rik; 24p3 receptor; 24p3R; AU041908; AW555662; BOCT; Boit; brain-specific organic ion transporter; brain-type organic cation transporter; hBOIT; Lipocalin-2 receptor; mBOCT; Neutrophil gelatinase-associated lipocalin receptor; NGAL receptor; NgalR; NGALR2; NGALR3; organic cation transporter BOCT; potent brain type organic ion transporter; rBOCT; SLC22A17; solute carrier family 22 (organic cation transporter), member 17; solute carrier family 22 member 17; solute carrier family 22, member 17 |
| Gene Symbols | SLC22A17 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?