missing translation for 'onlineSavingsMsg'
Learn More

SLC22A17 Antibody, DyLight 650, Novus Biologicals™

Product Code. 18429093 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429093 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429093 Supplier Novus Biologicals Supplier No. NBP176918C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC22A17 Polyclonal antibody specifically detects SLC22A17 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A17
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias 24p3R, BOCTNGALR3, BOIThBOIT, Brain-type organic cation transporter, Lipocalin-2 receptor, Neutrophil gelatinase-associated lipocalin receptor, NgalR, NGALR2, NGALRNGAL receptor, potent brain type organic ion transporter, solute carrier family 22 (organic cation transporter), member 17, solute carrier family 22 member 17,24p3 receptor, solute carrier family 22, member 17
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of the human Slc22A17. The immunogen is located within the last 50 amino acids of Slc22A17. Amino Acid Squence: CRRPSLLRQPPPTR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51310
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.