missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC22A16 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15837693
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15837693 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15837693 Supplier Invitrogen™ Supplier No. PA569281

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

This gene encodes a member of the organic zwitterion transporter protein family which transports carnitine. The encoded protein has also been shown to transport anticancer drugs like bleomycin successful treatment has been correlated with the level of activity of this transporter in tumor cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A16
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC22A16
Gene Accession No. Q86VW1
Gene Alias Carnitine transporter 2; CT2; dJ261K5.1; epididymis secretory protein Li 18; flipt 2; FLIPT2; fly-like putative organic ion transporter 2; fly-like putative transporter 2; HEL-S-18; OAT6; OCT6; OKB1; organic cation transporter 6; organic cation transporter OKB1; Organic cation/carnitine transporter 6; SLC22A16; solute carrier family 22 (organic cation transporter), member 16; solute carrier family 22 (organic cation/carnitine transporter), member 16; solute carrier family 22 member 16; WUGSC:RG331P03.1
Gene Symbols SLC22A16
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human SLC22A16.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85413
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.