missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A16 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-55400
This item is not returnable.
View return policy
Description
SLC22A16 Polyclonal specifically detects SLC22A16 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| SLC22A16 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Carnitine transporter 2, CT2Organic cation/carnitine transporter 6, Flipt 2, FLIPT2member 16, fly-like putative organic ion transporter 2, Fly-like putative transporter 2, organic cation transporter 6, Organic cation transporter OKB1, solute carrier family 22 (organic cation/carnitine transporter), member 16, solute carrier family 22 member 16 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 85413 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Immunohistochemistry 1-2ug/ml, Immunohistochemistry-Paraffin 1-2ug/ml, Peptide ELISA Detection limit 1:128000 | |
| NP_149116.2 | |
| SLC22A16 | |
| Peptide with sequence CSRNKRENTSSL corresponding to internal region according to NP_149116.2. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog, Pig and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction