missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99381-50ul
This item is not returnable.
View return policy
Description
SLC22A12 Polyclonal antibody specifically detects SLC22A12 in Human samples. It is validated for Western Blot
Specifications
| SLC22A12 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| OATL4, Organic anion transporter 4-like protein, Renal-specific transporter, RSTmember 12, solute carrier family 22 (organic anion/urate transporter), member 12, URAT1, URAT1Urate anion exchanger 1, urate transporter 1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SLC22A12. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 116085 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction