missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLC22A12 Polyclonal specifically detects SLC22A12 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | SLC22A12 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Alias | OATL4, Organic anion transporter 4-like protein, Renal-specific transporter, RSTmember 12, solute carrier family 22 (organic anion/urate transporter), member 12, URAT1, URAT1Urate anion exchanger 1, urate transporter 1 |
| Gene Symbols | SLC22A12 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?