missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SLC20A2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30703
This item is not returnable.
View return policy
Description
SLC20A2 Polyclonal antibody specifically detects SLC20A2 in Human,Mouse samples. It is validated for Western Blot
Specifications
| SLC20A2 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6575 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Gibbon ape leukemia virus receptor 2, Glvr-2, GLVR2PIT-2, hPit2, MLVAR, murine leukemia virus, amphotropic, receptor for, Phosphate transporter 2, pit2, PiT-2, sodium-dependent phosphate transporter 2, solute carrier family 20 (phosphate transporter), member 2, Solute carrier family 20 member 2 | |
| Between 300 and 350 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction