missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC1A7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550572
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total SLC1A7 protein.
This gene belongs to the sodium:dicarboxylate (SDF) symporter (TC 2.A.23) family, SLC1A7 subfamily. As transportor of L-glutamate; the L-glutamate uptake is sodium- and voltage-dependent and chloride-independent. Its associated chloride conductance may participate in visual processing.It interacts with the PDZ domains of DLG4. Expressed primarily in retina. Detectable in liver, heart, muscle and brain.
Specifications
| SLC1A7 | |
| Polyclonal | |
| Unconjugated | |
| Slc1a7 | |
| A930031E15Rik; AAAT; EAAT5; Excitatory amino acid transporter 5; excitatory amino acid transporter 5 (retinal glutamate transporter); retinal glutamate transporter; SLC1A7; solute carrier family 1 (glutamate transporter), member 7; solute carrier family 1 member 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6512 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O00341 | |
| Slc1a7 | |
| Synthetic peptide corresponding to a region derived from internal residues of human solute carrier family 1 (glutamate transporter), member 7. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction