missing translation for 'onlineSavingsMsg'
Learn More

SLC1A7 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18410586 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410586 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410586 Supplier Novus Biologicals Supplier No. NBP281802UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC1A7 Polyclonal antibody specifically detects SLC1A7 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC1A7
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias AAAT, EAAT5, excitatory amino acid transporter 5 (retinal glutamate transporter), excitatory amino acid transporter 5a long, retinal glutamate transporter, solute carrier family 1 (glutamate transporter), member 7
Host Species Rabbit
Immunogen SLC1A7 antibody was raised against a 14 amino acid peptide near the carboxy terminus of human SLC1A7. The immunogen is located within amino acids 460 - 510 of SLC1A7. Amino Acid Squence: RKDFARDTGTEKL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6512
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.