missing translation for 'onlineSavingsMsg'
Learn More

SLC1A5/ASCT2 Mouse, anti-Human, Clone: 1C4E1, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen SLC1A5/ASCT2
Clone 1C4E1
Concentration 1000 μg/mL
Applications Western Blot, Immunohistochemistry, ELISA
Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18096875
View Documents
Proteintech
68540-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18084485
View Documents
Proteintech
68540-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The SLC1A5/ASCT2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SLC1A5/ASCT2. This antibody recognizes human antigen. The SLC1A5/ASCT2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Specifications

SLC1A5/ASCT2
1000 μg/mL
Liquid
Human
AAAT, ASCT2, ATB(0), ATBO, Baboon M7 virus receptor, FLJ31068, M7V1, M7VS1, R16, RDR, RDRC, SLC1A5
Primary
1C4E1
Western Blot, Immunohistochemistry, ELISA
Mouse
6510
IgG1
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.