missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC1A4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576693
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Transporter for alanine, serine, cysteine, and threonine. Exhibits sodium dependence.
Specifications
| SLC1A4 | |
| Polyclonal | |
| Unconjugated | |
| SLC1A4 | |
| Alanine/serine/cysteine/threonine transporter 1; ASCT1; ASCT-1; AW045657; glutamate/neutral amino acid transporter; neutral amino acid transporter A; neutral amino acid transporter ASCT1; OTTHUMP00000159933; OTTHUMP00000235138; SATT; si:zc101n13.5; Slc1a4; slc1a4 protein; sodium-dependent neutral amino acid transporter ASCT1; solute carrier family 1 (glutamate/neutral amino acid transporter), member 4; solute carrier family 1 member 4; SPATCCM; zgc:92817 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 305540, 55963, 6509 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O35874, P43007 | |
| SLC1A4 | |
| Recombinant full length Human SLC1A4. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction