missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC19A3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £255.00
Specifications
| Antigen | SLC19A3 |
|---|---|
| Dilution | Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613082
|
Novus Biologicals
NBP2-97676-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18699654
|
Novus Biologicals
NBP2-97676-50ul |
50 μg |
£178.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC19A3 Polyclonal antibody specifically detects SLC19A3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SLC19A3 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Solute carrier family 19 member 3, solute carrier family 19, member 3, thiamine transporter 2, ThTr2, ThTr-2 | |
| Produced in rabbits immunized with E. coli-derived Human SLC19A3 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.0) | |
| 80704 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title