missing translation for 'onlineSavingsMsg'
Learn More

SLC19A2 Antibody (5B10), Novus Biologicals™

Product Code. 18349838 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349838 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349838 Supplier Novus Biologicals Supplier No. H00010560M10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SLC19A2 Monoclonal antibody specifically detects SLC19A2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC19A2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 5B10
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_008927.1
Gene Alias high affinity thiamine transporter, reduced folate carrier protein (RFC) like, solute carrier family 19 (thiamine transporter), member 2, Solute carrier family 19 member 2, TC1, Thiamine carrier 1, thiamine transporter 1, THT1, ThTr1, ThTr-1, THTR1thTr-1, TRMA
Host Species Mouse
Immunogen SLC19A2 (NP_008927.1, 209 a.a. ~ 285 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PMPQKSLFFHHIPSTCQRVNGIKVQNGGIVTDTPASNHLPGWEDIESKIPLNMEEPPVEEPEPKPDRLLVLKVLWND
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10560
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.