Learn More
Invitrogen™ SLC19A1 Polyclonal Antibody

Chicken Polyclonal Antibody
Brand: Invitrogen™ PA19553
Description
PA1-9553 detects SLC19A1 in human samples. PA1-9553 has been successfully used in Western blot procedures. By Western blot, PA1-9553 detects a 65 kDa protein representing SLC19A1. PA1-9553 immunogen is a purified recombinant SLC19A1 corresponding to the chromosome 21 gene (Human).
Transport of folate compounds into mammalian cells can occur via receptor-mediated or carrier-mediated mechanisms. A functional coordination between these 2 mechanisms has been proposed to be the method of folate uptake in certain cell types. Methotrexate (MTX) is an antifolate chemotherapeutic agent that is actively transported by the carrier-mediated uptake system. RFC1 plays a role in maintaining intracellular concentrations of folate.
Specifications
| SLC19A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC19A1 | |
| AI323572; CHMD; FLOT1; folate transporter 1; FOLT; IFC1; IFC-1; Intestinal folate carrier 1; intestinal folate carrier protein; methotrexate carrier 1; methotrexate carrier 2; methotrexate carrier 5; methotrexate carrier 6; MTX1; MTX-1; Placental folate transporter; reduced folate carrier 1; Reduced folate carrier protein; Reduced folate transporter; Reduced folate transporter 1; REFC; RFC; RFC1; RFC-1; RFT-1; SLC19A1; solute carrier family 19 (folate transporter), member 1; solute carrier family 19 (sodium/hydrogen exchanger), member 1; solute carrier family 19 member 1; solute carrier family 19, member 1 | |
| Chicken | |
| IgY fraction | |
| RUO | |
| 6573 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P41440 | |
| SLC19A1 | |
| Purified recombinant SLC19A1 corresponding to the chromosome 21 gene (Human). | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgY |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.