missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC19A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588778
This item is not returnable.
View return policy
Description
Immunogen sequence: LDGLRHCQRG HHPRQPPAQG LRSAAEEKAA QALSVQDKGL GGLQPAQSPP LSPEDSLGAV GPASLEQRQS DPYLAQAPAP QAAEFLSPVT TPSPCTLCSA QASGPEAADE TCPQLAVHPP GVSKLGLQCL PSDGVQNVNQ; Positive Samples: HeLa, U-87MG; Cellular Location: Membrane, Multi-pass membrane protein.
Transport of folate compounds into mammalian cells can occur via receptor-mediated or carrier-mediated mechanisms. A functional coordination between these 2 mechanisms has been proposed to be the method of folate uptake in certain cell types. Methotrexate (MTX) is an antifolate chemotherapeutic agent that is actively transported by the carrier-mediated uptake system. RFC1 plays a role in maintaining intracellular concentrations of folate.
Specifications
| SLC19A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC19A1 | |
| AI323572; CHMD; FLOT1; folate transporter 1; FOLT; IFC1; IFC-1; Intestinal folate carrier 1; intestinal folate carrier protein; methotrexate carrier 1; methotrexate carrier 2; methotrexate carrier 5; methotrexate carrier 6; MTX1; MTX-1; Placental folate transporter; reduced folate carrier 1; Reduced folate carrier protein; Reduced folate transporter; Reduced folate transporter 1; REFC; RFC; RFC1; RFC-1; RFT-1; SLC19A1; solute carrier family 19 (folate transporter), member 1; solute carrier family 19 (sodium/hydrogen exchanger), member 1; solute carrier family 19 member 1; solute carrier family 19, member 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6573 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P41440 | |
| SLC19A1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 452-591 of human SLC19A1 (NP_9192311). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction