missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC18B1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | C6orf192 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18245274
|
Novus Biologicals
NBP2-56822 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18690659
|
Novus Biologicals
NBP2-56822-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC18B1 Polyclonal specifically detects SLC18B1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| C6orf192 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| C6orf192, chromosome 6 open reading frame 192, dJ55C23.6, MFS-type transporter C6orf192, solute carrier family 18, subfamily B, member 1 | |
| SLC18B1 | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 116843 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MEALGDLEGPRAPGGDDPAGSAGETPGWLSR | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title