missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC18A3 Mouse anti-Human, Rat, HRP, Clone:S6-38, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of human SLC18A3.
Brand: Abnova MAB16987.100ug
This item is not returnable.
View return policy
Description
This gene is a member of the vesicular amine transporter family. The encoded transmembrane protein transports acetylcholine into secretory vesicles for release into the extracellular space. Acetylcholine transport utilizes a proton gradient established by a vacuolar ATPase. This gene is located within the first intron of the choline acetyltransferase gene. [provided by RefSeq]Specifications
| SLC18A3 | |
| Monoclonal | |
| HRP | |
| Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
| SLC18A3 | |
| Mouse | |
| 100 μg | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| S6-38 | |
| solute carrier family 18 (vesicular acetylcholine), member 3 | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| MGC12716, VACHT | |
| A synthetic peptide corresponding to amino acids 521-532 of human SLC18A3. | |
| 6572 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction