missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC18A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566633
This item is not returnable.
View return policy
Description
Immunogen sequence: KEEKLAILSQD CPMETRMYAT QKPTKEFPLG EDSDEEPDHE E Highest antigen sequence identity to the following orthologs - mouse 69%, rat 69%.
SLC18A1 (Solute Carrier Family 18 Member A1) is a Protein Coding gene. Diseases associated with SLC18A1 include Barbiturate Abuse and Adrenal Neuroblastoma. Among its related pathways are Transport of inorganic cations/anions and amino acids/oligopeptides and Synaptic vesicle pathway. Gene Ontology (GO) annotations related to this gene include monoamine transmembrane transporter activity and xenobiotic transmembrane transporter activity. An important paralog of this gene is SLC18A2.
Specifications
| SLC18A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC18A1 | |
| CGAT; VMAT | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6570 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P54219 | |
| SLC18A1 | |
| Recombinant Human SLC18A1. Recombinant protein control fragment (Product #RP-107286). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction