missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC17A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542138
This item is not returnable.
View return policy
Description
Peptide sequence: FVVIYDDPVS YPWISTSEKE YIISSLKQQV GSSKQPLPIK AMLRSLPIWS Sequence homology: Cow: 85%; Horse: 85%; Human: 100%.
SLC17A3 may be involved in actively transporting phosphate into cells via Na(+) cotransport.
Specifications
| SLC17A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC17A3 | |
| GOUT4; Na(+)/PI cotransporter 4; NPT4; SLC17A3; sodium/phosphate cotransporter 4; Sodium-dependent phosphate transport protein 4; solute carrier family 17 (organic anion transporter), member 3; solute carrier family 17 (sodium phosphate), member 3; solute carrier family 17 member 3; UAQTL4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10786 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O00476 | |
| SLC17A3 | |
| Synthetic peptide directed towards the middle region of human SLC17A3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction