missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC17A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568625
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 79%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.
SLC17A2 gene ontology annotations related to this gene include transmembrane transport.
Specifications
| SLC17A2 | |
| Polyclonal | |
| Unconjugated | |
| SLC17A2 | |
| C730032N17Rik; Na(+)/PI cotransporter 3; Npt3; SLC17A2; sodium phosphate transporter 3; sodium/phosphate cotransporter 3; sodium-dependent phosphate transport protein 3; sodium-dependent phosphate transport protein 3-like; solute carrier family 17 (sodium phosphate), member 2; solute carrier family 17 (vesicular glutamate transporter), member 2; solute carrier family 17 member 2; Solute carrier family 17 member 3; solute carrier family 17, member 2; solute carrier family 34 (sodium phosphate), member 1; Unknown (protein for MGC:134070) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 218103 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5SZA1 | |
| SLC17A2 | |
| synthetic peptide directed towards the middle region of human SLC17A2 (aa 281-330). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction