Learn More
Invitrogen™ SLC16A9 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA598506
Product Code. 16531332
  • £432.00 / 100µg
Description
SLC16A9 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Predicted to enable monocarboxylic acid transmembrane transporter activity. Involved in urate metabolic process. Predicted to be integral component of plasma membrane.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC16A9
Polyclonal
Unconjugated
SLC16A9
1200003C15Rik; 4930425B13Rik; AI552636; C10orf36; I79_016926; MCT 9; MCT9; monocarboxylate transporter; Monocarboxylate transporter 9; monocarboxylic acid transporter 9; Slc16a9; solute carrier family 16 (monocarboxylic acid transporters), member 9; solute carrier family 16 member 9; solute carrier family 16, member 9; solute carrier family 16, member 9 (monocarboxylic acid transporter 9)
Rabbit
Protein G
RUO
220963, 66859
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q7RTY1, Q7TM99
SLC16A9
Recombinant Human Monocarboxylate transporter 9 protein (185-304AA).
100 μg
Primary
Human, Mouse