missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC16A9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103927
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MOT9.
Predicted to enable monocarboxylic acid transmembrane transporter activity. Involved in urate metabolic process. Predicted to be integral component of plasma membrane.
Specifications
| SLC16A9 | |
| Polyclonal | |
| Unconjugated | |
| SLC16A9 | |
| 1200003C15Rik; 4930425B13Rik; AI552636; C10orf36; I79_016926; MCT 9; MCT9; monocarboxylate transporter; Monocarboxylate transporter 9; monocarboxylic acid transporter 9; Slc16a9; solute carrier family 16 (monocarboxylic acid transporters), member 9; solute carrier family 16 member 9; solute carrier family 16, member 9; solute carrier family 16, member 9 (monocarboxylic acid transporter 9) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 220963 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q7RTY1 | |
| SLC16A9 | |
| A synthesized peptide derived from human SLC16A9(Accession Q7RTY1), corresponding to amino acid residues Y165-N215. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction