missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC16A6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103926
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MOT7.
Proton-linked monocarboxylate transporter. Catalyzes the rapid transport across the plasma membrane of many monocarboxylates such as lactate, pyruvate, branched-chain oxo acids derived from leucine, valine and isoleucine, and the ketone bodies acetoacetate, beta-hydroxybutyrate and acetate.
Specifications
| SLC16A6 | |
| Polyclonal | |
| Unconjugated | |
| SLC16A6 | |
| AW743111; breast cancer protein BCY5; EGK_08910; ESTM12; MCT 6; MCT 7; MCT6; MCT7; Mct7B; monocarboxylate transporter; Monocarboxylate transporter 6; monocarboxylate transporter 7; monocarboxylate transporter 7B; monocarboxylate transporter 7-like; MOT7; Slc16a6; solute carrier family 16 (monocarboxylic acid transporters), member 6; solute carrier family 16 member 6; solute carrier family 16, member 6; solute carrier family 16, member 6 (monocarboxylic acid transporter 7) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 104681, 303772, 9120 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| B1AT66, O15403 | |
| SLC16A6 | |
| A synthesized peptide derived from human SLC16A6(Accession O15403), corresponding to amino acid residues G301-L351. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction