missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC16A6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569267
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 79%; Dog: 86%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 93%.
Proton-linked monocarboxylate transporter. Catalyzes the rapid transport across the plasma membrane of many monocarboxylates such as lactate, pyruvate, branched-chain oxo acids derived from leucine, valine and isoleucine, and the ketone bodies acetoacetate, beta-hydroxybutyrate and acetate.
Specifications
| SLC16A6 | |
| Polyclonal | |
| Unconjugated | |
| SLC16A6 | |
| AW743111; breast cancer protein BCY5; EGK_08910; ESTM12; MCT 6; MCT 7; MCT6; MCT7; Mct7B; monocarboxylate transporter; Monocarboxylate transporter 6; monocarboxylate transporter 7; monocarboxylate transporter 7B; monocarboxylate transporter 7-like; MOT7; Slc16a6; solute carrier family 16 (monocarboxylic acid transporters), member 6; solute carrier family 16 member 6; solute carrier family 16, member 6; solute carrier family 16, member 6 (monocarboxylic acid transporter 7) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9120 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O15403 | |
| SLC16A6 | |
| synthetic peptide directed towards the middle region of human SLC16A6. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction