missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC16A4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116599
This item is not returnable.
View return policy
Description
Positive Samples: SW480 Immunogen sequence: TFPLLMTYTI CFAIFAGGYL ALILPVLVDL CRNSTVNRFL GLASFFAGMA VLSGPPIAGW LYDYTQTYNG SFYFSGICYL LSSVSFFFVP LAERWKNSLT.
Proton-linked monocarboxylate transporter. Catalyzes the rapid transport across the plasma membrane of many monocarboxylates such as lactate, pyruvate, branched-chain oxo acids derived from leucine, valine and isoleucine, and the ketone bodies acetoacetate, beta-hydroxybutyrate and acetate.
Specifications
| SLC16A4 | |
| Polyclonal | |
| Unconjugated | |
| SLC16A4 | |
| AW146050; MCT 4; MCT 5; MCT4; MCT5; monocarboxylate transporter; monocarboxylate transporter 4; Monocarboxylate transporter 5; Slc16a4; solute carrier family 16 (monocarboxylic acid transporters), member 4; solute carrier family 16 member 4; solute carrier family 16, member 4; solute carrier family 16, member 4 (monocarboxylic acid transporter 5) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 295356, 9122 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15374 | |
| SLC16A4 | |
| A synthetic peptide corresponding to a sequence within amino acids 388-487 of human SLC16A4 (NP_0046871). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction