missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC16A4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98155-50ul
This item is not returnable.
View return policy
Description
SLC16A4 Polyclonal antibody specifically detects SLC16A4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SLC16A4 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| MCT 5, MCT 6, MCT6, Monocarboxylate transporter 5, Monocarboxylic acid transporter 5, Monocarboxylic acid transporter 6, MOT5, SLC16A5, solute carrier family 16 (monocarboxylic acid transporters) member 5, Solute carrier family 16 member 5 | |
| Produced in rabbits immunized with E. coli-derived Human SLC16A4 fragment. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 9122 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction