missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: AEEEKLHKPP ADSGVDLREV EHFLKAEPEK NGEVVHTPET SV Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.
Lactic acid and pyruvate transport across plasma membranes is catalyzed by members of the proton-linked monocarboxylate transporter (MCT) family, which has been designated solute carrier family-16. Each MCT appears to have slightly different substrate and inhibitor specificities and transport kinetics, which are related to the metabolic requirements of the tissues in which it is found. The MCTs, which include MCT1 (SLC16A1; MIM 600682) and MCT2 (SLC16A7; MIM 603654), are characterized by 12 predicted transmembrane domains (Price et al., 1998 [PubMed 9425115]).
Specifications
Specifications
| Antigen | SLC16A3 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SLC16A3 |
| Gene Accession No. | O15427 |
| Gene Alias | MCT 3; MCT 4; MCT3; MCT-3; Mct4; MCT-4; MGC138472; MGC138474; monocarboxylate transporter; monocarboxylate transporter 3; Monocarboxylate transporter 4; MOT4; Slc16a3; solute carrier family 16 (monocarboxylate transporter), member 3; solute carrier family 16 (monocarboxylic acid transporters), member 3; solute carrier family 16 member 3; solute carrier family 16, member 3 (monocarboxylic acid transporter 4) |
| Gene Symbols | SLC16A3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
