missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC16A2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA536978

Product Code. 15574181

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody detects endogenous protein at a molecular weight of 63 kDa. Purity is >95% by SDS-PAGE.

Monocarboxylates, such as lactate and pyruvate, play an integral role in cellular metabolism. Lactic acid is produced in large quantities as a result of glycolysis, which provides the majority of ATP to cells under normal physiological conditions. However, accumulation of lactic acid leads to a decrease in intracellular pH and cessation of glycolysis. In order for glycolysis to continue at a high rate, lactic acid must be transported out of the cell. This transport process is carried out by a family of monocarboxylate transporters (MCTs), which function as proton symports and are stereoselective for L-lactate. The MCT family consists of at least eight members, MCT 1-8, which contain between 10-12 transmembrane-helical (TM) domains, with the amino and carboxy termini located in the cytoplasm. Defects in the gene encoding for MCT8, SLC16A2, can cause monocarboxylate transporter 8 deficiency (MCT8 deficiency), a defect in cellular hormone transport causing a severe form of X-linked psychomotor retardation and abnormal thyroid levels.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC16A2
Polyclonal
Unconjugated
Slc16a2
AHDS; AW105741; DXS128; DXS128E; MCT 7; MCT 8; MCT7; MCT8; monocarboxylate transporter; monocarboxylate transporter 7; Monocarboxylate transporter 8; Monocarboxylate transporter 8 (MCT 8) (X-linked PEST-containing transporter); MRX22; SLC16A2; solute carrier family 16 (monocarboxylic acid transporters), member 2; solute carrier family 16 member 2; solute carrier family 16, member 2 (monocarboxylic acid transporter 8); solute carrier family 16, member 2 (thyroid hormone transporter); X-linked PEST-containing transporter; XPCT
Rabbit
Antigen affinity chromatography
RUO
20502, 259248, 6567
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
O70324, P36021, Q8K1P8
Slc16a2
Synthetic peptide corresponding to amino acids 152-197 of human SLC16A2.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.