missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC16A14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106682
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MOT14.
Proton-linked monocarboxylate transporter. May catalyze the transport of monocarboxylates across the plasma membrane.
Specifications
| SLC16A14 | |
| Polyclonal | |
| Unconjugated | |
| SLC16A14 | |
| 1110004H10Rik; MCT 14; MCT14; monocarboxylate transporter; Monocarboxylate transporter 14; monocarboxylic acid transporter 14; MOT14; Slc16a14; solute carrier family 16 (monocarboxylic acid transporters), member 14; solute carrier family 16 member 14; solute carrier family 16, member 14 (monocarboxylic acid transporter 14) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 151473, 71781 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q7RTX9, Q8K1C7 | |
| SLC16A14 | |
| A synthesized peptide derived from human SLC16A14(Accession Q7RTX9), corresponding to amino acid residues F130-K180. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction