missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC16A14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£231.00 - £484.00
Specifications
| Antigen | SLC16A14 |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18430261
|
Novus Biologicals
NBP1-83542-25ul |
25 μL |
£231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18275378
|
Novus Biologicals
NBP1-83542 |
0.1 mL |
£484.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC16A14 Polyclonal specifically detects SLC16A14 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SLC16A14 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 151473 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:YTSHEDIGYDFEDGPKDKKTLKPHPNIDGG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| MCT 14, MCT14member 14, monocarboxylate transporter 14, Solute carrier family 16 member 14, solute carrier family 16, member 14 (monocarboxylic acid transporter 14) | |
| SLC16A14 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title