missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC16A12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599651
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MCT12.
This gene encodes a transmembrane transporter that likely plays a role in monocarboxylic acid transport. A mutation in this gene has been associated with juvenile cataracts with microcornea and renal glucosuria.
Specifications
| SLC16A12 | |
| Polyclonal | |
| Unconjugated | |
| Slc16a12 | |
| 3230401A21; AW210596; CJMG; Creatine transporter 2; CRT2; LOW QUALITY PROTEIN: monocarboxylate transporter 12; MCT 12; MCT12; monocarboxylate transporter; Monocarboxylate transporter 12; monocarboxylic acid transporter 12; MOT12; RGD1311468; SLC16A12; solute carrier family 16 (monocarboxylic acid transporters), member 12; solute carrier family 16 member 12; solute carrier family 16, member 12; solute carrier family 16, member 12 (monocarboxylic acid transporter 12) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 387700 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6ZSM3 | |
| Slc16a12 | |
| A synthesized peptide derived from human SLC16A12(Accession Q6ZSM3), corresponding to amino acid residues I444-T486. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction