missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC16A11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568229
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
Proton-linked monocarboxylate transporter. Catalyzes the rapid transport across the plasma membrane of many monocarboxylates.
Specifications
| SLC16A11 | |
| Polyclonal | |
| Unconjugated | |
| SLC16A11 | |
| MCT11, MCT 11, monocarboxylate transporter 11, solute carrier family 16 (monocarboxylic acid transporters), member 11, solute carrier family 16 member 11 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 162515, 216867, 287450 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8NCK7, Q5NC32 | |
| SLC16A11 | |
| Synthesized peptide derived from human SLC16A11. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction