missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC16A10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550760
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total SLC16A10 protein.
Sodium-independent transporter that mediates the update of aromatic acid. Can function as a net efflux pathway for aromatic amino acids in the basosolateral epithelial cells.
Specifications
| SLC16A10 | |
| Polyclonal | |
| Unconjugated | |
| SLC16A10 | |
| 2610103N14Rik; 9830169E08; aromatic amino acid transporter 1; MCT 10; MCT10; monocarboxylate transporter; Monocarboxylate transporter 10; PRO0813; Slc16a10; solute carrier family 16 (aromatic amino acid transporter), member 10; solute carrier family 16 (monocarboxylic acid transporters), member 10; solute carrier family 16 member 10; solute carrier family 16, member 10 (aromatic amino acid transporter); TAT1; T-type amino acid transporter 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 117247, 72472 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1.4 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q3U9N9, Q8TF71 | |
| SLC16A10 | |
| Synthetic peptide corresponding to residues near the C terminal of human solute carrier family 16 (aromatic amino acid transporter), member 10. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction