missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC16A10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80700
This item is not returnable.
View return policy
Description
SLC16A10 Polyclonal specifically detects SLC16A10 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SLC16A10 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Q8TF71 | |
| SLC16A10 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SKKQREISKTTGKEKMEKMLENQNSLLSSSSGMFKKESDSI | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| member 10, solute carrier family 16, member 10 (aromatic amino acid transporter) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 117247 | |
| Human | |
| IgG |
Product Content Correction
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu