missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC15A3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566097
This item is not returnable.
View return policy
Description
Immunogen sequence: QSTYVLQGLHL HIPNIFPANP ANISVALRAQ GSSYTIPE Highest antigen sequence identity to the following orthologs - mouse 74%, rat 74%.
Proton oligopeptide cotransporter. Transports free histidine and certain di- and tripeptides.
Specifications
| SLC15A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC15A3 | |
| Solute carrier family 15 member 3; Osteoclast transporter; Peptide transporter 3; Peptide/histidine transporter 2; SLC15A3; OCTP; PHT2; PTR3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51296 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IY34 | |
| SLC15A3 | |
| Recombit Human SLC15A3 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction