missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC15A3 Polyclonal Antibody

SLC15A3 Polyclonal Antibody
Brand: Thermo Scientific PA561614
This item is not returnable.
View return policy
Description
SLC15A3 Polyclonal Antibody for IHC
Specifications
| SLC15A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC15A3 | |
| cAMP inducible gene 1; cAMP-inducible gene 1 protein; Ci1; cI-1; OCTP; Osteoclast transporter; peptide transporter 3; Peptide/histidine transporter 2; PHT2; PTR3; Slc15a3; solute carrier family 15 (oligopeptide transporter), member 3; solute carrier family 15 member 3; solute carrier family 15, member 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51296 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IY34 | |
| SLC15A3 | |
| Recombinant protein corresponding to Human SLC15A3. Recombinant protein control fragment (Product #RP-97303). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction