Learn More
SLC14A2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21932-1-AP-150UL
Description
The SLC14A2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC14A2. This antibody recognizes human, mouse antigen. The SLC14A2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| SLC14A2 | |
| 350 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| hUT A6, HUT2, SLC14A2, Urea transporter 2, Urea transporter, kidney, UT A2, UT2, UTA, UTR | |
| 150 μL | |
| 8170 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |