missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC13A4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570739
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 90%; Dog: 91%; Horse: 85%; Human: 100%; Mouse: 79%; Rat: 79%.
SLC13A4 gene ontology annotations related to this gene include sodium ion transport; transmembrane transport.
Specifications
| SLC13A4 | |
| Polyclonal | |
| Unconjugated | |
| Slc13a4 | |
| 9630060C05Rik; Na(+)/sulfate cotransporter SUT-1; NAS2; SLC13A4; sodium sulfate cotransporter-2; solute carrier family 13 (sodium/sulfate symporter), member 4; solute carrier family 13 (sodium/sulfate symporters), member 4; solute carrier family 13 (sodium/sulphate symporters), member 4; solute carrier family 13 member 4; sulphate transporter 1; SUT1; SUT-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26266 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UKG4 | |
| Slc13a4 | |
| synthetic peptide directed towards the middle region of human S13A4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction