missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC13A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553151
This item is not returnable.
View return policy
Description
Immunogen sequence: DFKAPNTETE PLLTWKKAQE TVPWNIILLL GGGFAMAKGC EESGLSVWIG GQLHPLENVP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Mammalian sodium-dicarboxylate cotransporters transport succinate and other Krebs cycle intermediates. They fall into 2 categories based on their substrate affinity: low affinity and high affinity. Both the low- and high-affinity transporters play an important role in the handling of citrate by the kidneys. The protein encoded by this gene represents the high-affinity form. Alternatively spliced transcript variants encoding different isoforms have been found for this gene, although the full-length nature of some of them have not been characterized yet.
Specifications
| SLC13A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC13A3 | |
| hNaDC3; hypothetical protein zgc:77173; mNaDC3; Na(+)/dicarboxylate cotransporter 3; Nadc3; NaDC-3; rNaDC3; SDCT2; slc13a3; sodium-dependent high affinity dicarboxylate transporter 3; sodium-dependent high-affinity dicarboxylate transporter 2; sodium-dependent high-affinity dicarboxylate transporter 3; solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3; solute carrier family 13 member 3; zgc:77173 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64849 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WWT9 | |
| SLC13A3 | |
| Recombinant protein fragment of Human SLC13A3 (Product #RP-91164) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction