Learn More
SLC13A3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553224
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SLFGQKEVRK DPSQESEENT AAVRRNGLHT VPTEMQFLAS TEAKDHPGET EVPLDLPADS RKEDEYRRNI WKGFL Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 67%.
Mammalian sodium-dicarboxylate cotransporters transport succinate and other Krebs cycle intermediates. They fall into 2 categories based on their substrate affinity: low affinity and high affinity. Both the low- and high-affinity transporters play an important role in the handling of citrate by the kidneys. The protein encoded by this gene represents the high-affinity form. Alternatively spliced transcript variants encoding different isoforms have been found for this gene, although the full-length nature of some of them have not been characterized yet.Specifications
SLC13A3 | |
Polyclonal | |
Unconjugated | |
SLC13A3 | |
hNaDC3; hypothetical protein zgc:77173; mNaDC3; Na(+)/dicarboxylate cotransporter 3; Nadc3; NaDC-3; rNaDC3; SDCT2; slc13a3; sodium-dependent high affinity dicarboxylate transporter 3; sodium-dependent high-affinity dicarboxylate transporter 2; sodium-dependent high-affinity dicarboxylate transporter 3; solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3; solute carrier family 13 member 3; zgc:77173 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64849 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WWT9 | |
SLC13A3 | |
Recombinant protein corresponding to Human SLC13A3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only