missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC13A3 Polyclonal Antibody

SLC13A3 Polyclonal Antibody
Brand: Invitrogen PA5110375
This item is not returnable.
View return policy
Description
SLC13A3 Polyclonal Antibody for Western Blot
Specifications
| SLC13A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC13A3 | |
| hNaDC3; hypothetical protein zgc:77173; mNaDC3; Na(+)/dicarboxylate cotransporter 3; Nadc3; NaDC-3; rNaDC3; SDCT2; slc13a3; sodium-dependent high affinity dicarboxylate transporter 3; sodium-dependent high-affinity dicarboxylate transporter 2; sodium-dependent high-affinity dicarboxylate transporter 3; solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3; solute carrier family 13 member 3; zgc:77173 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 64849 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.45 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8WWT9 | |
| SLC13A3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 160-240 of human SLC13A3 (NP_073740.2). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction