missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLC13A3/NaDC3 Polyclonal antibody specifically detects SLC13A3/NaDC3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | SLC13A3/NaDC3 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulation | PBS (pH 7.2) and 40% Glycerol |
| Gene Alias | hNaDC3, Na(+)/dicarboxylate cotransporter 3, NaDC-3, NADC3SDCT2naDC-3, sodium-dependent high affinity dicarboxylate transporter 3, Sodium-dependent high-affinity dicarboxylate transporter 2, solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3, solute carrier family 13 member 3 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: SLFGQKEVRKDPSQESEENTAAVRRNGLHTVPTEMQFLASTEAKDHPGETEVPLDLPADSRKEDEYRRNIWKGFL |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
¿Detecta una oportunidad de mejora?